Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBLN7
Entrez Gene ID
129804
Gene Name
fibulin 7
Gene Aliases
TM14
Chromosome Location
Chr.2: 112138385 - 112254888 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128165.1 NP_001121637.1 5 - 6 942 109
NM_153214.2 NP_694946.2 6 - 7 1080 109
XM_006712260.3 XP_006712323.1 6 - 7 1080 109
XM_011510585.2 XP_011508887.1 6 - 7 1080 109
XM_011510587.2 XP_011508889.1 7 - 8 1057 109
XM_017003317.1 XP_016858806.1 6 - 7 1080 109
XM_017003318.1 XP_016858807.1 6 - 7 1080 109
XM_017003319.1 XP_016858808.1 6 - 7 1080 109
GenBank mRNA AK094759.1 - 6 - 7 1080 109
AK124067.1 - 4 - 5 477 109
BC035784.1 - 6 - 7 1033 109
BC038242.1 - 2 - 3 275 109
BC126986.1 - 5 - 6 744 109

View Full Product Details