The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039083.3 | NP_001034172.3 | 1 - 2 | 88 | 147 |
NM_001103154.1 | NP_001096624.1 | 1 - 2 | 88 | 147 | |
XM_005256918.4 | XP_005256975.1 | 1 - 2 | 192 | 147 | |
XM_011524160.2 | XP_011522462.1 | 1 - 2 | 188 | 147 | |
XM_011524161.2 | XP_011522463.1 | 1 - 2 | 189 | 147 | |
XM_011524162.2 | XP_011522464.1 | 1 - 2 | 189 | 147 | |
XM_011524163.2 | XP_011522465.1 | 1 - 2 | 189 | 147 | |
XM_011524164.2 | XP_011522466.1 | 1 - 2 | 185 | 147 | |
XM_011524165.2 | XP_011522467.1 | 1 - 2 | 190 | 147 | |
XM_011524166.2 | XP_011522468.1 | 1 - 2 | 188 | 147 | |
XM_011524167.2 | XP_011522469.1 | 1 - 2 | 187 | 147 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113738.1 | NP_001107210.1 | 1 - 2 | 117 | 147 |
NM_001288811.1 | NP_001275740.1 | 1 - 2 | 117 | 147 | |
NM_001288812.1 | NP_001275741.1 | 1 - 2 | 117 | 147 | |
NM_001288813.1 | NP_001275742.1 | 1 - 2 | 117 | 147 | |
NM_016632.2 | NP_057716.2 | 1 - 2 | 117 | 147 | |
XM_005257439.4 | XP_005257496.1 | 1 - 2 | 176 | 147 | |
XM_011524875.2 | XP_011523177.1 | 1 - 2 | 170 | 147 | |
XM_011524876.2 | XP_011523178.1 | 1 - 2 | 169 | 147 | |
XM_011524877.2 | XP_011523179.1 | 1 - 2 | 165 | 147 | |
XM_011524878.2 | XP_011523180.1 | 1 - 2 | 167 | 147 | |
XM_017024729.1 | XP_016880218.1 | 1 - 2 | 169 | 147 | |
GenBank mRNA | DB065209.1 | - | 1 - 2 | 117 | 147 |
DB241677.1 | - | 1 - 2 | 97 | 147 |