Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXO41
Entrez Gene ID
150726
Gene Name
F-box protein 41
Gene Aliases
FBX41
Chromosome Location
Chr.2: 73254682 - 73284453 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080410.2 NP_001073879.2 5 - 6 1802 142
XM_005264158.3 XP_005264215.1 3 - 4 721 142
XM_006711950.2 XP_006712013.1 10 - 11 2786 142
XM_011532562.1 XP_011530864.1 6 - 7 2234 142
XM_011532563.2 XP_011530865.1 4 - 5 1159 142
XM_017003421.1 XP_016858910.1 7 - 8 4883 142
XM_017003422.1 XP_016858911.1 10 - 11 4528 142
XM_017003423.1 XP_016858912.1 10 - 11 3909 142
GenBank mRNA AB075820.1 - 5 - 6 1640 142
AK127503.1 - 2 - 3 283 142
BC112202.1 - 4 - 5 710 142

View Full Product Details