Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXO41
Entrez Gene ID
150726
Gene Name
F-box protein 41
Gene Aliases
FBX41
Chromosome Location
Chr.2: 73254682 - 73284453 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080410.2 NP_001073879.2 6 - 7 1919 127
XM_005264158.3 XP_005264215.1 4 - 5 838 127
XM_006711950.2 XP_006712013.1 11 - 12 2903 127
XM_011532562.1 XP_011530864.1 7 - 8 2351 127
XM_011532563.2 XP_011530865.1 5 - 6 1276 127
XM_017003421.1 XP_016858910.1 8 - 9 5000 127
XM_017003422.1 XP_016858911.1 11 - 12 4645 127
XM_017003423.1 XP_016858912.1 11 - 12 4026 127
GenBank mRNA AB075820.1 - 6 - 7 1757 127
AK127503.1 - 3 - 4 400 127
BC112202.1 - 5 - 6 827 127

View Full Product Details