Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXO41
Entrez Gene ID
150726
Gene Name
F-box protein 41
Gene Aliases
FBX41
Chromosome Location
Chr.2: 73254682 - 73284453 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080410.2 NP_001073879.2 9 - 10 2293 140
XM_005264158.3 XP_005264215.1 7 - 8 1212 140
XM_006711950.2 XP_006712013.1 14 - 15 3277 140
XM_011532562.1 XP_011530864.1 10 - 11 2725 140
XM_011532563.2 XP_011530865.1 8 - 9 1650 140
XM_017003421.1 XP_016858910.1 11 - 12 5374 140
XM_017003422.1 XP_016858911.1 14 - 15 5019 140
XM_017003423.1 XP_016858912.1 14 - 15 4400 140
GenBank mRNA AB075820.1 - 9 - 10 2131 140
AK127503.1 - 6 - 7 774 140
BC112202.1 - 8 - 9 1201 140

View Full Product Details