The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194272.1 | NP_919248.1 | 5 - 6 | 664 | 149 |
GenBank mRNA | AK124123.1 | - | 5 - 6 | 412 | 149 |
AK127873.1 | - | 5 - 6 | 680 | 149 | |
AY369207.1 | - | 5 - 6 | 664 | 149 | |
BC131612.1 | - | 4 - 5 | 316 | 149 | |
BC148208.1 | - | 4 - 5 | 316 | 149 |