The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019101.2 | NP_061974.2 | 1 - 2 | 189 | 153 |
XM_006715150.3 | XP_006715213.1 | 1 - 2 | 166 | 153 | |
GenBank mRNA | AF118393.3 | - | 1 - 2 | 149 | 153 |
AJ245434.1 | - | 1 - 2 | 165 | 153 | |
BC020683.1 | - | 1 - 2 | 168 | 153 | |
BG564719.1 | - | 1 - 2 | 190 | 153 |