Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APOM
Entrez Gene ID
55937
Gene Name
apolipoprotein M
Gene Aliases
G3a, HSPC336, NG20, apo-M
Chromosome Location
Chr.6: 31652410 - 31658210 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256169.1 NP_001243098.1 4 - 5 465 141
NM_019101.2 NP_061974.2 4 - 5 510 141
NR_045828.1 4 - 5 472 141
XM_006715150.3 XP_006715213.1 4 - 5 494 141
GenBank mRNA AA846749.1 - 2 - 3 141
AF118393.3 - 4 - 5 470 141
AF161454.1 - 4 - 5 431 141
AJ245434.1 - 4 - 5 486 141
BC020683.1 - 4 - 5 489 141
BF344944.1 - 4 - 5 444 140
CB054016.1 - 4 - 5 472 141
CN428415.1 - 4 - 5 452 141

View Full Product Details