Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MXD4
Entrez Gene ID
10608
Gene Name
MAX dimerization protein 4
Gene Aliases
MAD4, MST149, MSTP149, bHLHc12
Chromosome Location
Chr.4: 2247432 - 2262143 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017007656.1 XP_016863145.1 2 - 3 275 120

View Full Product Details