Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LRRC37BP1
- Entrez Gene ID
-
147172
- Gene Name
- leucine rich repeat containing 37B pseudogene 1
- Gene Aliases
-
LRRC37B2
- Chromosome Location
-
Chr.17: 30631755 - 30637477 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK124001.1 |
- |
2 - 2 |
1902 |
156 |
AK124759.1 |
- |
2 - 2 |
2309 |
156 |
AL713754.1 |
- |
2 - 2 |
1427 |
156 |
- Gene Symbol
- LOC107133515
- Entrez Gene ID
-
107133515
- Gene Name
- SMURF2P1-LRRC37BP1 readthrough transcribed pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 30576465 - 30637466 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_015341.2 |
|
9 - 9 |
2164 |
156 |
GenBank mRNA |
AK075282.1 |
- |
6 - 6 |
1440 |
156 |
View Full Product Details