The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172089.3 | NP_742086.1 | 7 - 8 | 676 | 143 |
GenBank mRNA | AY081051.1 | - | 7 - 8 | 635 | 143 |