The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017975.4 | NP_001017975.4 | 33 - 34 | 3772 | 77 |
XM_011540849.1 | XP_011539151.1 | 33 - 34 | 3855 | 77 | |
XM_011540850.2 | XP_011539152.1 | 33 - 34 | 3784 | 77 | |
XM_011540851.1 | XP_011539153.1 | 33 - 34 | 3739 | 77 | |
XM_011540852.2 | XP_011539154.1 | 33 - 34 | 3721 | 77 | |
XM_011540855.1 | XP_011539157.1 | 31 - 32 | 3577 | 77 | |
XM_011540857.1 | XP_011539159.1 | 31 - 32 | 3336 | 77 | |
XM_011540859.2 | XP_011539161.1 | 30 - 31 | 3305 | 77 | |
XM_017000490.1 | XP_016855979.1 | 32 - 33 | 3635 | 77 | |
XM_017000491.1 | XP_016855980.1 | 32 - 33 | 3584 | 77 | |
XM_017000492.1 | XP_016855981.1 | 27 - 28 | 2819 | 77 | |
XM_017000493.1 | XP_016855982.1 | 21 - 22 | 2023 | 77 | |
GenBank mRNA | AB044787.1 | - | 10 - 11 | 1060 | 77 |
AB074274.1 | - | 5 - 6 | 523 | 77 | |
AB204867.1 | - | 33 - 34 | 3773 | 77 | |
AK094079.1 | - | 16 - 17 | 1599 | 77 | |
AK294754.1 | - | 27 - 28 | 2790 | 77 | |
AK302582.1 | - | 29 - 30 | 2940 | 77 | |
BC130525.1 | - | 18 - 19 | 1718 | 77 | |
BC144172.1 | - | 18 - 19 | 1718 | 77 | |
BX649037.1 | - | 14 - 15 | 1308 | 77 |