The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017975.4 | NP_001017975.4 | 26 - 27 | 2993 | 171 |
XM_011540849.1 | XP_011539151.1 | 26 - 27 | 3076 | 171 | |
XM_011540850.2 | XP_011539152.1 | 26 - 27 | 3005 | 171 | |
XM_011540851.1 | XP_011539153.1 | 26 - 27 | 2960 | 171 | |
XM_011540852.2 | XP_011539154.1 | 26 - 27 | 2942 | 171 | |
XM_011540855.1 | XP_011539157.1 | 24 - 25 | 2798 | 171 | |
XM_011540857.1 | XP_011539159.1 | 24 - 25 | 2557 | 171 | |
XM_011540859.2 | XP_011539161.1 | 23 - 24 | 2526 | 171 | |
XM_017000490.1 | XP_016855979.1 | 25 - 26 | 2856 | 171 | |
XM_017000492.1 | XP_016855981.1 | 20 - 21 | 2040 | 171 | |
XM_017000493.1 | XP_016855982.1 | 14 - 15 | 1244 | 171 | |
GenBank mRNA | AB044787.1 | - | 3 - 4 | 281 | 171 |
AB204867.1 | - | 26 - 27 | 2994 | 171 | |
AK094079.1 | - | 9 - 10 | 820 | 171 | |
AK294754.1 | - | 20 - 21 | 2010 | 171 | |
AK302582.1 | - | 22 - 23 | 2161 | 171 | |
BC130525.1 | - | 11 - 12 | 939 | 171 | |
BC144172.1 | - | 11 - 12 | 939 | 171 |