The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201286.3 | NP_958443.1 | 2 - 2 | 1715 | 73 |
XM_017029299.1 | XP_016884788.1 | 3 - 3 | 1982 | 73 | |
XM_017029300.1 | XP_016884789.1 | 3 - 3 | 1865 | 73 | |
XM_017029301.1 | XP_016884790.1 | 2 - 2 | 1785 | 73 | |
GenBank mRNA | AJ583823.2 | - | 1 - 1 | 1636 | 73 |