Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP4V2
Entrez Gene ID
285440
Gene Name
cytochrome P450 family 4 subfamily V member 2
Gene Aliases
BCD, CYP4AH1
Chromosome Location
Chr.4: 186191520 - 186213463 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207352.3 NP_997235.3 3 - 4 719 88
XM_005262935.3 XP_005262992.1 3 - 4 504 88
GenBank mRNA AK122600.1 - 3 - 4 454 88
AK126473.1 - 4 - 5 388 88
AY422002.1 - 3 - 4 719 88
BC060857.1 - 3 - 4 585 88

View Full Product Details