Product Details
- Assay Reference Genome
Location
Chr.14:88416728 on build GRCh38
- Cytoband
- 14q31.3
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 55812
- Gene Symbol
- SPATA7
- Gene Name
- spermatogenesis associated 7
- Gene Aliases
- HEL-S-296, HSD-3.1, HSD3, LCA3
- Location
- Chr.14:88385638-88470350 on build GRCh38
- Assay Gene Location
- Overlaps Intron 5 - Exon 6
View Full Product Details