The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_015404.1 | 2 - 2 | 2203 | 118 | |
GenBank mRNA | AK096935.1 | - | 2 - 2 | 2203 | 118 |
AK124158.1 | - | 1 - 1 | 2073 | 118 | |
BC007973.2 | - | 3 - 3 | 521 | 118 | |
BC015633.2 | - | 3 - 3 | 890 | 118 | |
BC023547.2 | - | 2 - 2 | 1854 | 118 | |
BC107695.1 | - | 2 - 2 | 774 | 118 | |
JQ794500.1 | - | 2 - 2 | 2306 | 118 |