The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166395.1 | NP_001159867.1 | 2 - 2 | 1962 | 140 |
NM_005769.2 | NP_005760.1 | 2 - 2 | 1910 | 140 | |
GenBank mRNA | AF131235.1 | - | 2 - 2 | 1880 | 140 |
AF280088.1 | - | 2 - 2 | 1820 | 140 | |
AK026635.1 | - | 2 - 2 | 1770 | 140 | |
AK074746.1 | - | 2 - 2 | 1767 | 140 | |
BC035282.1 | - | 2 - 2 | 1776 | 140 |