Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TET2
Entrez Gene ID
54790
Gene Name
tet methylcytosine dioxygenase 2
Gene Aliases
KIAA1546, MDS
Chromosome Location
Chr.4: 105145875 - 105279803 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017628.4 NP_060098.3 3 - 3 9146 159
GenBank mRNA BC150180.1 - 3 - 3 4853 159
BC150201.1 - 3 - 3 4853 159
BX640738.1 - 3 - 3 9157 159

View Full Product Details