The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020786.2 | NP_065837.1 | 2 - 2 | 3323 | 131 |
XM_005256064.4 | XP_005256121.1 | 2 - 2 | 3370 | 131 | |
XM_005256066.4 | XP_005256123.1 | 2 - 2 | 3412 | 131 | |
XM_005256067.4 | XP_005256124.1 | 2 - 2 | 3437 | 131 | |
XM_005256068.4 | XP_005256125.1 | 3 - 3 | 3384 | 131 | |
XM_017023506.1 | XP_016878995.1 | 3 - 3 | 3518 | 131 | |
GenBank mRNA | AB037769.1 | - | 2 - 2 | 3270 | 131 |