The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032367.2 | NP_115743.1 | 3 - 3 | 1254 | 125 |
GenBank mRNA | BC007239.1 | - | 3 - 3 | 1254 | 125 |