Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR6K2
Entrez Gene ID
81448
Gene Name
olfactory receptor family 6 subfamily K member 2
Gene Aliases
OR1-17
Chromosome Location
Chr.1: 158699678 - 158700652 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005279.1 NP_001005279.1 1 - 1 845 164
GenBank mRNA BC137022.1 - 1 - 1 889 164
BC137023.1 - 1 - 1 930 164

View Full Product Details