Product Details

Assay Reference Genome
Location

Chr.9:35079771 on build GRCh38
Cytoband
9p13.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2189
Gene Symbol
FANCG
Gene Name
Fanconi anemia complementation group G
Gene Aliases
FAG, XRCC9
Location
Chr.9:35073838-35080016 on build GRCh38
Assay Gene Location
Within Exon 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
FANCG NM_004629.1 1 246 NP_004620.1
AJ007669.1 1 246 CAA07602.1
AK293427.1
AK312987.1 1 171
BC011623.2 AAH11623.1
KU159422.1 1 277

View Full Product Details