Product Details

Assay Reference Genome
Location

Chr.8:38413331 on build GRCh38
Cytoband
8p11.23
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2260
Gene Symbol
FGFR1
Gene Name
fibroblast growth factor receptor 1
Gene Aliases
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
Location
Chr.8:38411138-38468834 on build GRCh38
Assay Gene Location
Within Exon 23
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
FGFR1 NM_001174063.1 18 3702 NP_001167534.1
NM_001174064.1 19 3770 NP_001167535.1
NM_001174065.1 18 3022 NP_001167536.1
NM_001174066.1 17 2761 NP_001167537.1
NM_001174067.1 19 3182 NP_001167538.1
NM_015850.3 18 3702 NP_056934.2
NM_023105.2 17 3441 NP_075593.1
NM_023106.2 17 3435 NP_075594.1
NM_023110.2 18 3708 NP_075598.2
XM_006716303.2 XP_006716366.1
XM_006716304.1 XP_006716367.1
XM_006716306.2 XP_006716369.1
XM_006716307.1 XP_006716370.1
XM_006716309.3 XP_006716372.1
XM_006716310.2 XP_006716373.1
XM_006716311.1 XP_006716374.1
XM_006716312.1 XP_006716375.1
XM_006716313.2 XP_006716376.1
XM_006716314.1 XP_006716377.1
XM_011544443.2 XP_011542745.1
XM_011544444.1 XP_011542746.1
XM_011544445.2 XP_011542747.1
XM_011544446.2 18 3009 XP_011542748.1
XM_011544447.2 18 3003 XP_011542749.1
XM_011544448.1 XP_011542750.1
XM_011544449.1 XP_011542751.1
XM_011544450.2 17 2607 XP_011542752.1
XM_011544451.1 XP_011542753.1
XM_017013219.1 XP_016868708.1
XM_017013220.1 18 2997 XP_016868709.1
XM_017013221.1 XP_016868710.1
XM_017013222.1 XP_016868711.1
XM_017013223.1 XP_016868712.1
XM_017013224.1 18 3497 XP_016868713.1
XM_017013225.1 18 2980 XP_016868714.1
XM_017013226.1 17 2607 XP_016868715.1
XM_017013227.1 17 2601 XP_016868716.1
XM_017013228.1 17 3230 XP_016868717.1
XM_017013229.1 XP_016868718.1
XM_017013230.1 16 3795 XP_016868719.1
AB208919.1 19 3557 BAD92156.1
AK222718.1 18 3501 BAD96438.1
AK292470.1 17 2761
BC015035.1 19 3430 AAH15035.1
BC018128.1 18 3466 AAH18128.1
BC091494.1 16 2572 AAH91494.1
BX649167.1 14 5050
FJ809917.1 18 3717
M34185.1 17 2754 AAA35836.1
M34186.1 17 3019 AAA35837.1
M34641.1 17 2769 AAA35835.1
M37722.1 17 2721 AAA75007.1
M60485.1 19 3431 AAA35840.1
M63887.1 17 2766 AAA35958.1
M63888.1 17 2791 AAA35959.1
X66945.1 18 3492 CAA47375.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv951128 Chr8:38411783 - 38418282 on Build GRCh38 Deletion FGFR1

View Full Product Details