Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR6M1
Entrez Gene ID
390261
Gene Name
olfactory receptor family 6 subfamily M member 1
Gene Aliases
OR11-271
Chromosome Location
Chr.11: 123805408 - 123806349 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005325.1 NP_001005325.1 1 - 1 894 97
GenBank mRNA BC136929.1 - 1 - 1 932 97
BC136931.1 - 1 - 1 932 97

View Full Product Details