The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005492.1 | NP_001005492.1 | 1 - 1 | 527 | 142 |
GenBank mRNA | X64974.1 | - | 1 - 1 | 153 | 142 |