The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000689.4 | NP_000680.2 | 13 - 13 | 2214 | 149 |
GenBank mRNA | AB209821.1 | - | 13 - 13 | 1941 | 149 |
AI074764.1 | - | 1 - 1 | 149 | ||
AK000118.1 | - | 13 - 13 | 1941 | 149 | |
AK026641.1 | - | 13 - 13 | 1941 | 149 | |
EU668318.1 | - | 13 - 13 | 1941 | 149 | |
FJ224286.1 | - | 13 - 13 | 1950 | 149 | |
K03000.1 | - | 9 - 9 | 1415 | 149 |