The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039847.2 | NP_001034936.1 | 7 - 7 | 811 | 135 |
NM_001039848.2 | NP_001034937.1 | 7 - 7 | 867 | 135 | |
NM_002085.4 | NP_002076.2 | 7 - 7 | 789 | 135 | |
GenBank mRNA | AI274351.1 | - | 1 - 1 | 135 | |
AK225859.1 | - | 7 - 7 | 708 | 135 | |
AL558552.3 | - | 7 - 7 | 664 | 134 | |
BC010157.2 | - | 7 - 7 | 617 | 135 | |
BC011836.2 | - | 7 - 7 | 710 | 135 | |
BC021567.1 | - | 7 - 7 | 703 | 135 | |
BC022071.1 | - | 7 - 7 | 726 | 135 | |
BC032695.1 | - | 7 - 7 | 710 | 135 | |
BC039849.1 | - | 7 - 7 | 778 | 135 | |
BC046163.1 | - | 7 - 7 | 685 | 135 | |
BC058329.1 | - | 7 - 7 | 706 | 135 | |
BQ648717.1 | - | 7 - 7 | 654 | 135 | |
X71973.1 | - | 7 - 7 | 751 | 135 |