The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023944.3 | NP_076433.3 | 7 - 7 | 949 | 154 |
NR_117085.1 | 6 - 6 | 967 | 154 | ||
XM_006722850.3 | XP_006722913.1 | 7 - 7 | 834 | 154 | |
XM_011528202.2 | XP_011526504.1 | 5 - 5 | 469 | 154 | |
XM_011528203.2 | XP_011526505.1 | 4 - 4 | 1691 | 154 | |
XM_011528204.2 | XP_011526506.1 | 5 - 5 | 1170 | 154 | |
XM_011528205.2 | XP_011526507.1 | 4 - 4 | 382 | 154 | |
XM_011528207.2 | XP_011526509.1 | 4 - 4 | 2078 | 154 | |
XM_011528208.2 | XP_011526510.1 | 6 - 6 | 588 | 154 | |
XM_017027172.1 | XP_016882661.1 | 7 - 7 | 891 | 154 | |
GenBank mRNA | AB035130.1 | - | 7 - 7 | 860 | 154 |
AB035131.1 | - | 7 - 7 | 860 | 154 | |
AK074412.1 | - | 2 - 2 | 1197 | 154 | |
AK075435.1 | - | 6 - 6 | 895 | 154 | |
AL832171.1 | - | 7 - 7 | 802 | 154 | |
AY008841.1 | - | 6 - 6 | 830 | 154 | |
AY358977.1 | - | 7 - 7 | 871 | 154 | |
BC035350.1 | - | 7 - 7 | 876 | 154 |