Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLPS
Entrez Gene ID
1208
Gene Name
colipase
Gene Aliases
-
Chromosome Location
Chr.6: 35794982 - 35797344 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252597.1 NP_001239526.1 4 - 4 605 89
NM_001252598.1 NP_001239527.1 2 - 2 384 89
NM_001832.3 NP_001823.1 3 - 3 507 89
GenBank mRNA BC007061.1 - 3 - 3 486 89
BC017897.1 - 3 - 3 486 89
BP321569.1 - 2 - 2 361 89
BP322637.1 - 3 - 3 486 89
BP324927.1 - 3 - 3 486 89
J02883.1 - 3 - 3 472 89

View Full Product Details