The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166304.1 | NP_001159776.1 | 7 - 7 | 1098 | 102 |
NM_017861.3 | NP_060331.3 | 6 - 6 | 1044 | 102 | |
GenBank mRNA | AK294340.1 | - | 8 - 8 | 1398 | 102 |
BC022542.1 | - | 6 - 6 | 1004 | 102 |