The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144701.2 | NP_653302.2 | 11 - 11 | 2372 | 111 |
XM_005270516.2 | XP_005270573.1 | 8 - 8 | 2018 | 111 | |
GenBank mRNA | AF461422.1 | - | 11 - 11 | 2372 | 111 |
BC040720.2 | - | 4 - 4 | 1357 | 111 |