Product Details

Assay Reference Genome
Location

Chr.9:108881693 on build GRCh38
Cytoband
9q31.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
8518
Gene Symbol
IKBKAP
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
Gene Aliases
DYS, ELP1, FD, IKAP, IKI3, TOT1
Location
Chr.9:108867520-108934332 on build GRCh38
Assay Gene Location
Overlaps Intron 31 - Exon 31
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
IKBKAP NM_001318360.1 NP_001305289.1
NM_003640.4 NP_003631.2
XM_011519136.1 XP_011517438.1
XM_011519137.1 XP_011517439.1
XM_017015238.1 XP_016870727.1
AB210033.1
AF044195.1 AAC64258.1
AF153419.2 AAG43369.1
AK001641.1
AK022559.1
AK024223.1
AK123257.1
AK127237.1
AK225798.1
AK289962.1
AK304742.1
AL049945.1 CAB43219.1
BC033094.1 AAH33094.1
CR749385.1 CAH18235.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv6659 Chr9:108856263 - 108890282 on Build GRCh38 Insertion ACTL7A IKBKAP

View Full Product Details