Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHB
Entrez Gene ID
5245
Gene Name
prohibitin
Gene Aliases
HEL-215, HEL-S-54e, PHB1
Chromosome Location
Chr.17: 49404048 - 49414905 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281496.1 NP_001268425.1 7 - 7 1410 153
NM_001281497.1 NP_001268426.1 6 - 6 1011 153
NM_001281715.1 NP_001268644.1 8 - 8 1454 153
NM_002634.3 NP_002625.1 7 - 7 1362 153
XM_017024763.1 XP_016880252.1 8 - 8 1504 153
GenBank mRNA AK223181.1 - 7 - 7 1337 153
BM972128.1 - 1 - 1 153
BP267961.1 - 1 - 1 245 153
BQ574485.1 - 1 - 1 153

View Full Product Details