The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040135.2 | NP_001035225.1 | 9 - 10 | 1089 | 89 |
XM_017012464.1 | XP_016867953.1 | 8 - 9 | 1030 | 89 | |
XM_017012465.1 | XP_016867954.1 | 8 - 9 | 893 | 89 | |
GenBank mRNA | BC015207.1 | - | 9 - 10 | 1001 | 89 |