The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004559.4 | NP_004550.2 | 4 - 5 | 683 | 68 |
NR_132737.1 | 3 - 4 | 501 | 68 | ||
GenBank mRNA | AK309996.1 | - | 4 - 5 | 320 | 68 |
BC000064.2 | - | 4 - 5 | 470 | 68 | |
BC002411.2 | - | 4 - 5 | 466 | 68 | |
BC010430.1 | - | 4 - 5 | 465 | 68 | |
BC013838.1 | - | 3 - 4 | 501 | 68 | |
BC015208.1 | - | 4 - 5 | 415 | 68 | |
BC018393.1 | - | 4 - 5 | 450 | 68 | |
BC038384.1 | - | 4 - 5 | 472 | 68 | |
BC065571.1 | - | 4 - 5 | 490 | 68 | |
BC070084.1 | - | 4 - 5 | 490 | 68 | |
BC071708.1 | - | 4 - 5 | 475 | 68 | |
BC072014.1 | - | 4 - 5 | 475 | 68 | |
BC090038.1 | - | 4 - 5 | 475 | 68 | |
BC098435.1 | - | 4 - 5 | 481 | 68 | |
BC106045.1 | - | 4 - 5 | 477 | 68 | |
L28809.1 | - | 4 - 5 | 433 | 68 | |
M24070.1 | - | 4 - 5 | 472 | 68 | |
M83234.1 | - | 1 - 1 | 577 | 68 | |
M85234.1 | - | 1 - 1 | 583 | 68 |