Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB44
Entrez Gene ID
29068
Gene Name
zinc finger and BTB domain containing 44
Gene Aliases
BTBD15, HSPC063, ZNF851
Chromosome Location
Chr.11: 130226677 - 130314908 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014155.4 NP_054874.3 5 - 6 1742 86
XM_006718826.3 XP_006718889.1 5 - 6 1763 86
XM_017017624.1 XP_016873113.1 5 - 6 1760 86
GenBank mRNA BC071729.1 - 5 - 6 1721 86
CD671410.1 - 1 - 2 74 86
CN423489.1 - 2 - 3 167 86

View Full Product Details