Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
MKS1 | NM_001165927.1 | 4 | 386 | NP_001159399.1 |
NM_001321268.1 | 4 | 397 | NP_001308197.1 | |
NM_001321269.1 | 4 | 397 | NP_001308198.1 | |
NM_017777.3 | 4 | 397 | NP_060247.2 | |
XM_005257485.3 | 4 | 318 | XP_005257542.1 | |
XM_006721965.2 | 4 | 329 | XP_006722028.1 | |
XM_011524957.2 | 4 | 377 | XP_011523259.1 | |
XM_011524958.2 | 4 | 377 | XP_011523260.1 | |
XM_011524959.2 | 4 | 376 | XP_011523261.1 | |
XM_011524960.2 | 4 | 376 | XP_011523262.1 | |
XM_017024803.1 | 4 | 345 | XP_016880292.1 | |
XM_017024804.1 | 4 | 345 | XP_016880293.1 | |
XM_017024805.1 | 4 | 351 | XP_016880294.1 | |
AK000352.1 | 1 | 67 | BAA91105.1 | |
AK301020.1 | 4 | 328 | ||
AK310815.1 | 4 | 386 | ||
BC010061.2 | 4 | 332 | AAH10061.1 | |
DQ185029.1 | 4 | 397 | AAZ94714.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517229 | Chr17:58176226 - 58373163 on Build GRCh38 | Gain+Loss | MIR142 MIR4736 RNF43 EPX TSPOAP1 MPO MKS1 SUPT4H1 TSPOAP1-AS1 LPO |