Product Details

Assay Reference Genome
Location

Chr.9:27558547 on build GRCh38
Cytoband
9p21.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
203228
Gene Symbol
C9orf72
Gene Name
chromosome 9 open reading frame 72
Gene Aliases
ALSFTD, FTDALS, FTDALS1
Location
Chr.9:27546546-27573866 on build GRCh38
Assay Gene Location
Within Exon 8
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
C9orf72 NM_001256054.2 7 1001 NP_001242983.1
NM_018325.4 7 906 NP_060795.1
AK291425.1 7 896
AL832229.1 6 863
AL832467.1 6 2481
BC068445.1 6 855 AAH68445.1
JN681271.1 7 979

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1015259 Chr9:26347305 - 27643043 on Build GRCh38 Gain IFT74-AS1 IFNK EQTN C9orf72 LRRC19 LINC00032 TEK PLAA MOB3B IFT74 CAAP1

View Full Product Details