Product Details

Assay Reference Genome
Location

Chr.9:108869048 on build GRCh38
Cytoband
9q31.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
8518
Gene Symbol
IKBKAP
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
Gene Aliases
DYS, ELP1, FD, IKAP, IKI3, TOT1
Location
Chr.9:108867520-108934332 on build GRCh38
Assay Gene Location
Within Exon 37
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
IKBKAP NM_001318360.1 37 4445 NP_001305289.1
NM_003640.4 37 4590 NP_003631.2
XM_011519136.1 37 4424 XP_011517438.1
XM_011519137.1 37 4198 XP_011517439.1
XM_017015238.1 35 4161 XP_016870727.1
AB210033.1 37 4352
AF044195.1 37 4369 AAC64258.1
AF153419.2 37 4376 AAG43369.1
AK001641.1 37 4374
AK022559.1 19 2059
AK024223.1 17 1833
AK123257.1 10 2146
AK127237.1 29 4005
AK225798.1 24 2674
AK289962.1 37 4382
AK304742.1 37 4087
AL049945.1 12 1290 CAB43219.1
BC033094.1 37 4152 AAH33094.1
CR749385.1 30 3417 CAH18235.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv6659 Chr9:108856263 - 108890282 on Build GRCh38 Insertion IKBKAP ACTL7A

View Full Product Details