Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEZ6L2
Entrez Gene ID
26470
Gene Name
seizure related 6 homolog like 2
Gene Aliases
BSRPA, PSK-1
Chromosome Location
Chr.16: 29871159 - 29899573 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114100.2 NP_001107572.1 13 - 14 2424 81
NM_001243332.1 NP_001230261.1 15 - 16 2766 81
NM_001243333.1 NP_001230262.1 14 - 15 2634 81
NM_012410.3 NP_036542.1 15 - 16 2556 81
XM_017023135.1 XP_016878624.1 13 - 14 2523 81
GenBank mRNA AB105376.1 - 15 - 16 2395 81
AF131749.1 - 7 - 8 1287 81
AJ245820.1 - 15 - 16 2395 81
AJ245821.1 - 15 - 16 2395 81
AK027407.1 - 13 - 14 2036 81
AK027676.1 - 12 - 13 2006 81
AK074998.1 - 11 - 12 1724 81
AK293675.1 - 14 - 15 2479 81
AK299103.1 - 15 - 16 2763 81
BC000567.2 - 13 - 14 2224 81
KJ534938.1 - 12 - 13 1990 81
KJ534940.1 - 14 - 15 2320 81
KJ534941.1 - 13 - 14 2179 81
KJ535092.1 - 15 - 16 2520 81

View Full Product Details