Product Details

Assay Reference Genome
Location

Chr.8:38415969 on build GRCh38
Cytoband
8p11.23
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2260
Gene Symbol
FGFR1
Gene Name
fibroblast growth factor receptor 1
Gene Aliases
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
Location
Chr.8:38411138-38468834 on build GRCh38
Assay Gene Location
Within Exon 18
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
FGFR1 NM_001174063.1 13 2691 NP_001167534.1
NM_001174064.1 14 2759 NP_001167535.1
NM_001174065.1 13 2011 NP_001167536.1
NM_001174066.1 12 1750 NP_001167537.1
NM_001174067.1 14 2171 NP_001167538.1
NM_015850.3 13 2691 NP_056934.2
NM_023105.2 12 2430 NP_075593.1
NM_023106.2 12 2424 NP_075594.1
NM_023110.2 13 2697 NP_075598.2
XM_006716303.2 13 2498 XP_006716366.1
XM_006716304.1 13 2014 XP_006716367.1
XM_006716306.2 13 2492 XP_006716369.1
XM_006716307.1 13 2008 XP_006716370.1
XM_006716309.3 13 2468 XP_006716372.1
XM_006716310.2 12 2231 XP_006716373.1
XM_006716311.1 12 1629 XP_006716374.1
XM_006716312.1 12 1699 XP_006716375.1
XM_006716313.2 12 2225 XP_006716376.1
XM_006716314.1 12 1703 XP_006716377.1
XM_011544443.2 13 2003 XP_011542745.1
XM_011544444.1 13 1992 XP_011542746.1
XM_011544445.2 13 2005 XP_011542747.1
XM_011544446.2 13 1998 XP_011542748.1
XM_011544447.2 13 1992 XP_011542749.1
XM_011544448.1 12 1602 XP_011542750.1
XM_011544449.1 12 1596 XP_011542751.1
XM_011544450.2 12 1596 XP_011542752.1
XM_011544451.1 11 1465 XP_011542753.1
XM_017013219.1 13 1993 XP_016868708.1
XM_017013220.1 13 1986 XP_016868709.1
XM_017013221.1 14 2207 XP_016868710.1
XM_017013222.1 13 2492 XP_016868711.1
XM_017013223.1 13 2002 XP_016868712.1
XM_017013224.1 13 2486 XP_016868713.1
XM_017013225.1 13 1969 XP_016868714.1
XM_017013226.1 12 1596 XP_016868715.1
XM_017013227.1 12 1590 XP_016868716.1
XM_017013228.1 12 2219 XP_016868717.1
XM_017013229.1 7 2178 XP_016868718.1
XM_017013230.1 11 2784 XP_016868719.1
AB208919.1 14 2546 BAD92156.1
AK222718.1 13 2490 BAD96438.1
AK291754.1 13 1890
AK292470.1 12 1750
AK309947.1 14 2050
BC015035.1 14 2419 AAH15035.1
BC018128.1 13 2455 AAH18128.1
BC091494.1 11 1561 AAH91494.1
BX649167.1 9 4039
FJ809917.1 13 2706
KU178066.1 12 1749
KU178067.1 11 1492
KU178068.1 11 1486
KU178069.1 11 1482
KU178070.1 11 1476
M34185.1 12 1743 AAA35836.1
M34186.1 12 2008 AAA35837.1
M34641.1 12 1758 AAA35835.1
M37722.1 12 1710 AAA75007.1
M60485.1 14 2420 AAA35840.1
M63887.1 12 1755 AAA35958.1
M63888.1 12 1755 AAA35959.1
M63889.1 AAA35960.1
X51803.1 13 1872 CAA36101.1
X52833.1 12 1842 CAA37015.1
X57119.1 12 1650 CAA40401.1
X57120.1 13 1911 CAA40402.1
X57121.1 13 1917 CAA40403.1
X57122.1 12 1644 CAA40404.1
X66945.1 13 2481 CAA47375.1
Y00665.1 9 1155 CAA68679.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv951128 Chr8:38411783 - 38418282 on Build GRCh38 Deletion FGFR1

View Full Product Details