Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
PINK1 | NM_032409.2 | 2 | 629 | NP_115785.1 |
AB053323.1 | 2 | 629 | BAB55647.1 | |
AF316873.1 | 2 | 605 | AAK28062.1 | |
BC028215.1 | 2 | 544 | AAH28215.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv545731 | Chr1:19989106 - 20776306 on Build GRCh38 | Gain | CAMK2N1 KIF17 DDOST UBXN10 PLA2G2F FAM43B LINC01141 HP1BP3 PLA2G2D MIR6084 SH2D5 UBXN10-AS1 MUL1 VWA5B1 PINK1 PINK1-AS CDA PLA2G5 PLA2G2C |
nsv834369 | Chr1:20609770 - 20779837 on Build GRCh38 | Loss | KIF17 DDOST PINK1 PINK1-AS HP1BP3 CDA MIR6084 SH2D5 |