The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144658.3 | NP_653259.3 | 34 - 35 | 3839 | 77 |
XM_005262368.3 | XP_005262425.1 | 35 - 36 | 4123 | 77 | |
XM_005262369.4 | XP_005262426.1 | 35 - 36 | 3817 | 77 | |
XM_011531276.2 | XP_011529578.1 | 35 - 36 | 3758 | 77 | |
XM_011531278.2 | XP_011529580.1 | 22 - 23 | 2244 | 77 | |
GenBank mRNA | AK056684.1 | - | 4 - 5 | 353 | 77 |
AK124735.1 | - | 29 - 30 | 3262 | 77 | |
AK125641.1 | - | 13 - 14 | 1351 | 77 | |
AY692226.1 | - | 34 - 35 | 3839 | 77 |