The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194434.2 | NP_919415.2 | 4 - 5 | 721 | 156 |
XM_017026079.1 | XP_016881568.1 | 4 - 5 | 497 | 156 | |
GenBank mRNA | AF044670.1 | - | 4 - 5 | 425 | 156 |
AF057358.1 | - | 4 - 5 | 401 | 156 | |
AF086627.1 | - | 4 - 5 | 401 | 156 | |
AF154847.1 | - | 4 - 5 | 438 | 156 | |
AK292948.1 | - | 4 - 5 | 449 | 156 | |
AK315577.1 | - | 4 - 5 | 660 | 156 | |
BC002992.1 | - | 4 - 5 | 635 | 156 | |
BT019618.1 | - | 4 - 5 | 401 | 156 |