The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000683.3 | NP_000674.2 | 1 - 1 | 646 | 93 |
GenBank mRNA | BC142625.1 | - | 1 - 1 | 817 | 93 |
J03853.1 | - | 1 - 1 | 646 | 93 |