Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR1F2P
Entrez Gene ID
26184
Gene Name
olfactory receptor family 1 subfamily F member 2 pseudogene
Gene Aliases
OLFMF2, OR16-3, OR1F11, OR1F2, OR1F3P, hg91
Chromosome Location
Chr.16: 3215562 - 3216546 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_002169.1 1 - 1 50 73


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream olfactory receptor family 1 subfamily F member 1 OR1F1 OLFMF, OR16-36, OR16-37, OR16-88, OR16-89, OR16-90, OR1F10, OR1F13P, OR1F4, OR1F5, OR1F6, OR1F7, OR1F8, OR1F9, OR3-145, ORL1023 -10.2kb Sense Chr. - 3188195 - 3206559 on Build GRCh38
Downstream zinc finger protein 200 ZNF200 5.8kb Anti-sense Chr. - 3222325 - 3236235 on Build GRCh38

View Full Product Details