Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLEU2
Entrez Gene ID
8847
Gene Name
deleted in lymphocytic leukemia 2 (non-protein coding)
Gene Aliases
1B4, BCMSUN, DLB2, LEU2, LINC00022, MIR15AHG, NCRNA00022, RFP2OS, TRIM13OS
Chromosome Location
Chr.13: 49982552 - 50125541 on Build GRCh38
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location Amplicon Length
RefSeq NR_002612.1
7 - 8 1017 115
GenBank mRNA AY455757.1 - LIT3420
DLEU2
10 - 11 1317 115


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream microRNA 3613 MIR3613 -573.0 Sense Chr. - 49996415 - 49996501 on Build GRCh38
Downstream microRNA 16-1 MIR16-1 MIRN16-1, miRNA16-1, mir-16-1 21.6kb Sense Chr. - 50048973 - 50049061 on Build GRCh38

View Full Product Details