Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SNORD90
- Entrez Gene ID
-
692206
- Gene Name
- small nucleolar RNA, C/D box 90
- Gene Aliases
-
HBII-295
- Chromosome Location
-
Chr.9: 122880213 - 122880319 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003071.1 |
|
1 - 1 |
51 |
87 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ring finger and CCCH-type domains 2 |
RC3H2 |
MNAB, RNF164 |
|
Sense |
Chr. - 122849453 - 122905283 on Build GRCh38 |
Downstream |
zinc finger and BTB domain containing 6 |
ZBTB6 |
ZID, ZNF482 |
27.7kb
|
Sense |
Chr. - 122908056 - 122913328 on Build GRCh38 |
View Full Product Details