Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR2A42
Entrez Gene ID
402317
Gene Name
olfactory receptor family 2 subfamily A member 42
Gene Aliases
-
Chromosome Location
Chr.7: 144231911 - 144232843 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001802.2 NP_001001802.2 1 - 1 871 148

Gene Symbol
OR2A1
Entrez Gene ID
346528
Gene Name
olfactory receptor family 2 subfamily A member 1
Gene Aliases
-
Chromosome Location
Chr.7: 144318125 - 144319057 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005287.1 NP_001005287.1 1 - 1 871 148

View Full Product Details